Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HMGA2, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP151985

Catalog No. NBP151985


Add to cart

Description

Description

HMGA2 Polyclonal antibody specifically detects HMGA2 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

HMGA2
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
HMGA2
Peptide with sequence C-KAAQKKAEATGEK corresponding to internal region according to NP_003474.1, NP_003475.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize both reported isoforms (NP_003474.1; NP_003475.1) and is not expected to cross-react with HMGA1.
ELISA, Western Blot
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:8000
P52926
BABLHMGI-C, high mobility group AT-hook 2, High mobility group AT-hook protein 2, high mobility group protein HMGI-C, high-mobility group (nonhistone chromosomal) protein isoform I-C, High-mobility group protein HMGI-C, HMGICSTQTL9, LIPO
Goat
IgG
0.1 mg
Cancer
Primary
8091
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only