Learn More
HMGA2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA137871
Description
This gene encodes a protein that belongs to the non-histone chromosomal high mobility group (HMG) protein family. HMG proteins function as architectural factors and are essential components of the enhancesome. This protein contains structural DNA-binding domains and may act as a transcriptional regulating factor. Identification of the deletion, amplification, and rearrangement of this gene that are associated with myxoid liposarcoma suggests a role in adipogenesis and mesenchymal differentiation. A gene knock out study of the mouse counterpart demonstrated that this gene is involved in diet-induced obesity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is lung carcinoma.
Specifications
HMGA2 | |
Polyclonal | |
HMGA2 | |
BABL, HMGI-C, HMGIC, LIPO, STQTL9, High-mobility group protein HMGI-C, high mobility group AT-hook protein 2, high mobility group protein HMGI-C, high-mobility group nonhistone chromosomal protein isoform I-C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8091 | |
4° C, do not freeze | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
P52926 | |
HMGA2 | |
Synthetic peptide from C-terminus of human HMGA2. | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.