Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGA2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525276

Catalog No. PIPA525276


Add to cart

Description

Description

This antibody is predicted to react with bovine based on sequence homology.

This gene encodes a protein that belongs to the non-histone chromosomal high mobility group (HMG) protein family. HMG proteins function as architectural factors and are essential components of the enhancesome. This protein contains structural DNA-binding domains and may act as a transcriptional regulating factor. Identification of the deletion, amplification, and rearrangement of this gene that are associated with myxoid liposarcoma suggests a role in adipogenesis and mesenchymal differentiation. A gene knock out study of the mouse counterpart demonstrated that this gene is involved in diet-induced obesity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications

Specifications

HMGA2
Polyclonal
PBS with 0.09% sodium azide
P52926
BABL, HMGI-C, HMGIC, LIPO, STQTL9, High-mobility group protein HMGI-C, high mobility group AT-hook protein 2, high mobility group protein HMGI-C, high-mobility group (nonhistone chromosomal) protein isoform I-C
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8091
Flow Cytometry, Western Blot
Unconjugated
HMGA2
Liquid
HMGA2
KLH conjugated synthetic peptide between 64-92 amino acids from the C-terminal region of human HMGA2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit