Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGB1/HMG-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $409.50
Specifications
Antigen | HMGB1/HMG-1 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 5-10ug/ml∼, Simple Western 1:100, Flow Cytometry 0.5ug/10^6cells, Immunohistochemistry-Paraffin 5ug/ml |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227117SS
|
Novus Biologicals
NBP227117SS |
0.025 mg |
Each for $204.00
|
|
|||||
NBP227117
|
Novus Biologicals
NBP227117 |
0.1 mg |
Each for $409.50
|
|
|||||
Description
HMGB1/HMG-1 Polyclonal specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HMGB1/HMG-1 | |
Western Blot 5-10ug/ml∼, Simple Western 1:100, Flow Cytometry 0.5ug/10^6cells, Immunohistochemistry-Paraffin 5ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
P09429 | |
3146 | |
A portion of amino acids 100-166 of human HMGB1 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
HMGB1 | |
IgG | |
Protein A purified | |
The HMGB1 amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, chicken, cow and monkey. This antibody recognizes both B box domain (amino acids 88-162) and full length HMGB1 protein. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title