Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HMGB3/HMG4, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP126398

Catalog No. NBP126398

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

HMGB3/HMG4
0.5 mg/mL
Western Blot 0.01 - 0.03 ug/ml, Peptide ELISA Detection limit 1:64000
NP_005333.2
High mobility group protein 2a, High mobility group protein 4, high mobility group protein B3, high-mobility group (nonhistone chromosomal) protein 4, high-mobility group box 3, HMG-2a, HMG2AHMG-4, HMG4MGC90319, non-histone chromosomal protein
Goat
IgG
0.1 mg
Chromatin Research, DNA Repair
Primary
3149
Western Blot, ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
HMGB3
Peptide with sequence C-KFDGAKGPAKVARKK corresponding to internal region (near C-Terminus) according to NP_005333.2.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only