Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGB3/HMG4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | HMGB3/HMG4 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 3-5 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224645SS
|
Novus Biologicals
NBP224645SS |
0.025 mg |
Each for $204.00
|
N/A | |||||
NBP224645
|
Novus Biologicals
NBP224645 |
0.1 mg |
Each for $409.50
|
N/A | |||||
Description
HMGB3/HMG4 Polyclonal specifically detects HMGB3/HMG4 in Human, Mouse, Rat, Bovine, Equine, Opossum samples. It is validated for Western Blot.Specifications
HMGB3/HMG4 | |
Western Blot 3-5 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Bovine, Equine, Opossum | |
O15347 | |
3149 | |
A portion of amino acids 1-50 of human HMGB3 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Chromatin Research, DNA Repair | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
High mobility group protein 2a, High mobility group protein 4, high mobility group protein B3, high-mobility group (nonhistone chromosomal) protein 4, high-mobility group box 3, HMG-2a, HMG2AHMG-4, HMG4MGC90319, non-histone chromosomal protein | |
HMGB3 | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, chimpanzee, rhesus monkey and dog, 96% homologous in cow, 93% homologous inopossumand 80% homologous in chicken. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title