Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGB3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523361
Description
HMGB3 Polyclonal antibody specifically detects HMGB3 in Bovine, Equine, Human, Marsupial, Mouse, Rat samples. It is validated for Western Blot.Specifications
HMGB3 | |
Polyclonal | |
Unconjugated | |
HMGB3 | |
high mobility group box 3; high mobility group protein 1; high mobility group protein 2a; High mobility group protein 4; high mobility group protein B1; high mobility group protein B3; high mobilty group protein 2A; high-mobility group (nonhistone chromosomal) protein 4; high-mobility group protein 4; HMG-1; HMG2a; HMG-2a; HMG4; HMG-4; HMGB1; Hmgb3; HMGB3 transcript variant 1/2; MGC90319; RGD1564407; Unknown (protein for MGC:133786) | |
Rabbit | |
Protein A | |
RUO | |
100069284, 15354, 305373, 3149, 613729 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
O15347, O54879, Q32L31 | |
HMGB3 | |
A portion of amino acids 1-50 of human HMGB3 | |
100 μg | |
Primary | |
Bovine, Equine, Human, Marsupial, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction