Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HMGCR, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136961

Catalog No. NBP136961


Add to cart

Description

Description

HMG-CoA Reductase/HMGCR Polyclonal antibody specifically detects HMG-CoA Reductase/HMGCR in Human samples. It is validated for Peptide ELISA, Western Blot (Negative).
Specifications

Specifications

HMG-CoA Reductase/HMGCR
0.5 mg/mL
Peptide ELISA Detection limit 1:8000, Western Blot (Negative)
P04035
3-hydroxy-3-methylglutaryl CoA reductase (NADPH), 3-hydroxy-3-methylglutaryl-CoA reductase, 3-hydroxy-3-methylglutaryl-Coenzyme A reductase, EC 1.1.1, EC 1.1.1.34, HMG-CoA reductase, hydroxymethylglutaryl-CoA reductase, LDLCQ3
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody recognizes and epitope within the internal region. Expected from sequence similarity: Human.
ELISA, Western Blot
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
HMGCR
Peptide with sequence C-ADPSPQNSTADTSK corresponding to internal region according to NP_000850.1.
Immunogen affinity purified
RUO
Primary
3156
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only