Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522310

Catalog No. PIPA522310


Add to cart

Description

Description

PA5-22310 targets HNF-1 alpha in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-22310 immunogen is a recombit fragment corresponding to a region within amino acids 35 and 268 of HNF1 alpha (Uniprot IDNo.P20823). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

HNF1A
Polyclonal
Unconjugated
HNF1A
Liquid
HNF1A
Recombit fragment corresponding to a region within amino acids 35 and 268 of HNF1 alpha (Uniprot IDNo.P20823)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.47 mg/mL
PBS with 1% BSA, 20% glycerol and 0.025% Proclin 300; pH 7
P20823, P22361, P15257
HNF-1A, HNF1, IDDM20, LFB1, MODY3, TCF-1, TCF1, HNF-1-alpha, albumin proximal factor, hepatic nuclear factor 1, hepatocyte nuclear factor 1-alpha, interferon production regulator factor, liver-specific transcription factor LF-B1, transcription factor 1, hepatic
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
21405, 24817, 6927
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit