Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF6 Rabbit anti-Canine, Human, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523459

Catalog No. PIPA523459


Add to cart

Description

Description

Hepatocyte nuclear factor 6 is a CUT homeobox family protein which is a transcription factor and contains a CUT DNA-binding domain and a homeobox DNA-binding domain. It is an upstream regulator of several genes involved in the pathogenesis of maturity-onset diabetes of the young. HNF6 is a ligand of the FOXM1 transcription factor network. HNF6, along with HNF4, controls gene expression in the liver. The expression levels of HNF6 and HNF4 rise steadily during development and that up- or down regulation of HNF6 and HNF4 expression generate premature or delayed expression of the g6pc gene.
Specifications

Specifications

HNF6
Polyclonal
Unconjugated
ONECUT1
Liquid
ONECUT1
Synthetic peptide (439 RSLDKWQDEGSSNSGNS 455) of HNF6 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Canine, Human, Non-human primate
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q9UBC0
HNF-6, HNF6, HNF6A, hepatocyte nuclear factor 6, hepatocyte nuclear factor 6, alpha, one cut domain family member 1, one cut domain, family member 1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3175, 487558
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit