Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP A1 Mouse anti-Human, Clone: 4B10, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051521
Description
Use Anti-hnRNP A1 Antibody, clone 4B10 (mouse monoclonal antibody) validated in western blotting to detect hnRNP A1 also known as Helix-destabilizing protein, Single-strand RNA-binding protein, heterogeneous nuclear ribonucleoprotein A1.Specifications
hnRNP A1 | |
Monoclonal | |
Unconjugated | |
NP_002127.1 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG2b κ |
Western Blot | |
4B10 | |
Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
HNRNPA1, HNRPA1, hnRNPA1 | |
Full length native protein partially purified human hnRNP A1. | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only