Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP A2/B1 Mouse anti-Human, Clone: DP383, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051522
Description
Use Anti-hnRNP A2/B1 Antibody, clone DP383 (Mouse Monoclonal Antibody) validated in western blot, immunohistochemistry, immunoprecipitation to detect hnRNP A2/B1 also known as heterogeneous nuclear ribonucleoprotein A2/B1.Specifications
hnRNP A2/B1 | |
Monoclonal | |
Unconjugated | |
NP_002128 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG2a κ |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
DP383 | |
Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1, HNRPB1, RNPA2, SNRPB1, HNRNPA2B1 | |
Recombinant protein corresponding to human hnRNP A2/B1. | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only