Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hnRNP U Mouse anti-Human, Clone: 3G6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051516
Description
Use Anti-hnRNP U Antibody, clone 3G6 (mouse monoclonal antibody) validated in western blot, immunoprecipitation, immunocytochemistry, ELISA to detect hnRNP U also known as heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A).Specifications
hnRNP U | |
Monoclonal | |
Unconjugated | |
NP_004492.2 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG2b κ |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
3G6 | |
Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
hnrnpu, HNRPU, SAF-A, SAFA, U21.1, hnRNPU, p120, pp120 | |
Recombinant protein corresponding to human hnRNP U. | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only