Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hnRNP K Rabbit anti-Human, Marsupial, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523437

Catalog No. PIPA523437


Add to cart

Description

Description

hnRNPK is most extensively studied among heterogeneous nuclear ribonucleoproteins family, which plays an important role in cell signaling, telomere elongation, DNA repair and regulation of expression at both transcriptional and translational levels. It is a highly conserved RNA/DNA-binding protein which interacts with different classes of factors of extracellular signals.
Specifications

Specifications

hnRNP K
Polyclonal
Unconjugated
HNRNPK
Liquid
HNRNPK
Synthetic peptide (11 PNTETNGEFGKRPAEDMEE 29) of hnRNPK protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Marsupial, Mouse, Non-human primate, Rat
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P61978, P61979, P61980
RP11-575L7.1, CSBP, HNRPK, TUNP, dC-stretch binding protein, transformation upregulated nuclear protein
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
117282, 15387, 3190
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit