Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNRPA3 Rabbit anti-Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP180487

Catalog No. NBP180487


Add to cart

Description

Description

HNRPA3 Polyclonal antibody specifically detects HNRPA3 in Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

HNRPA3
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
HNRNPA3
Synthetic peptide directed towards the N terminal of human HNRPA3. Peptide sequence PGRPQPDSGRRRRRRGEEGHDPKEPEQLRKLFIGGLSFETTDDSLREHFE.
Protein A purified
RUO
Primary
220988
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
NP_919223
heterogeneous nuclear ribonucleoprotein A3
Rabbit
IgG
100ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Human: 100%.
Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Rabbit, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only