Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXA9 Rabbit anti-Chicken, Feline, Human, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523367

Catalog No. PIPA523367


Add to cart

Description

Description

Homeobox containing genes are a family of transcription factors regulating normal development and controlling primary cellular processes like cell identity, cell division and differentiation. The HOXA9 gene encodes a class I homeodomain protein which is involved in hematopoesis and myeloid differentiation. It shows different splice variants like a 1.8-kb homeobox-containing transcript in fetal brain, lung, liver, and kidney, and 2.2- and 3.3-kb transcripts in fetal and adult kidney and in adult skeletal muscle. Fusion between the HOXA9 gene and the nucleoporin gene NUP98, a member of the GLFG nucleoporin family, on 11p15 occurs as a consequence of leukemia-associated chromosomal translocations. So NUP98-HOXA9 is associated with the t(7;11)(p15;p15) translocation in acute myeloid leukemia, myelodysplastic syndrome, and blastic crisis of chronic myeloid leukemia.
Specifications

Specifications

HOXA9
Polyclonal
Unconjugated
HOXA9
Liquid
HOXA9
Synthetic peptide (256 NRRMKMKKINKDRAK 270) of Homeobox protein A9 protein.
Protein A
RUO
Antibody
Polyclonal
Chicken, Feline, Human, Mouse, Non-human primate, Rat
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q98924, P31269, P09631
ABD-B, HOX1, HOX1.7, HOX1G, homeobox protein Hox-1G, homeobox protein Hox-A9, homeodomain protein HOXA9
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
101101453, 15405, 297099, 3205, 396096
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit