Learn More
HOXB13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA141133
Catalog No. 01-675-713
Add to cart
Description
Description
HOXB13 is a transcription factor that belongs to the homeobox gene family. Genes of this family are highly conserved among vertebrates and essential for vertebrate embryonic development. This gene has been implicated to play a role in fetal skin development and cutaneous regeneration. In mice, a similar gene was shown to exhibit temporal and spatial colinearity in the main body axis of the embryo, but was not expressed in the secondary axes, which suggests functions in body patterning along the axis. This gene and other HOXB genes form a gene cluster at chromosome the 17q21-22 region.
Specifications
Specifications
HOXB13
Polyclonal
whole serum with 50% glycerol and 0.025% sodium azide
Q92826, P70321
HOXB13, HOXA13
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Western Blot
Unconjugated
HOXB13
Liquid
HOXB13
Synthetic peptide corresponding to residues E(232) R E Y A A N K F I T K D K R R K I(249) of human HOXB13.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10481, 15408, 303480