Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXD8 Rabbit anti-Human, Mouse, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523476

Catalog No. PIPA523476


Add to cart

Description

Description

Homeobox protein HOXD8 belongs to the Antp homeobox family and contains one homeobox DNA-binding domain. It is a sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis. HOXD8 has redundant functions at upper thoracic and sacral levels, including positioning of the hind limbs. It is reported that HOXD8 may have a role in scarless wound repair.
Specifications

Specifications

HOXD8
Polyclonal
Unconjugated
HOXD8
Liquid
HOXD8
Synthetic peptide (264 SRQEVKDGETKKEAQELE 281) of Homeobox protein D8 protein.
Protein A
RUO
Antibody
Polyclonal
Human, Mouse, Non-human primate
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P13378, P23463
HOX4, HOX4E, HOX5.4, Hox-4.5, homeo box 4E, homeo box D8, homeobox protein 5.4, homeobox protein Hox-4E, homeobox protein Hox-5.4, homeobox protein Hox-D8
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
15437, 3234
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit