Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOMER1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514188

Catalog No. PIPA514188

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

HOMER1 is a member of the homer family of dendritic proteins. Members of this family regulate group 1 metabotrophic glutamate receptor function.
Specifications

Specifications

HOMER1
Polyclonal
PBS with 0.09% sodium azide
Q86YM7
Homer protein homolog 1; SYN47; HOMER1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9456
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
HOMER1
Liquid
HOMER1
KLH conjugated synthetic peptide between 96-122 amino acids from the N-terminal region of human HOMER1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit