Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOMEZ Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524260

Catalog No. PIPA524260

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May function as a transcriptional regulator.
Specifications

Specifications

HOMEZ
Polyclonal
PBS with 0.09% sodium azide
Q8IX15, Q80W88
KIAA1443, homeobox and leucine zipper protein Homez, homeodomain leucine zipper-containing factor
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
239099, 57594
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
HOMEZ
Liquid
HOMEZ
KLH conjugated synthetic peptide between 158-186 amino acids from the N-terminal region of human HOMEZ
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit