Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSL Chicken anti-Mouse, Rat, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA19573

Catalog No. 01-674-449


Add to cart

Description

Description

PA1-9573 detects HSL in mouse samples. PA1-9573 has been successfully used in Western blot and ELISA procedures. By Western blot, PA1-9573 detects a 83 kDa protein representing HSL . PA1-9573 immunogen is a synthetic peptide based on mouse HSL protein.

Hormone-sensitive lipase (HSL) is an intracellular neutral lipase found in a variety of tissues, but primarily in adipose and steroidogenic tissues. It hydrolyzes triglycerides and cholesteryl esters. It is the major enzyme responsible for the mobilization of free fatty acids from adopise tissue.
Specifications

Specifications

HSL
Polyclonal
Unconjugated
Lipe
Liquid
LIPE
Synthetic peptide based on mouse HSL protein.
IgY fraction
RUO
Antibody
Polyclonal
Mouse, Rat
ELISA, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P54310, P15304
HSL antibody, hormone sensitive lipase antibody.
Chicken
IgY
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
16890, 25330
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit