Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Equine IgG Isotype Control, Agarose, Novus Biologicals™
Horse Antibody
Supplier: Novus Biologicals NBP197139
Description
IgG specifically detects IgG in Equine samples. It is validated for Immunoprecipitation.Specifications
IgG | |
Agarose | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: None. with 0.01% Sodium Azide | |
IGHG1 | |
Multi-step | |
RUO | |
3500 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunoprecipitation | |
Immunoprecipitation 1:10 - 1:500 | |
Immunoglobulin G, ImmunoglobulinG | |
Equine | |
10 mg | |
Isotype Control (Primary) | |
Equine | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction