Learn More
anti-Horse IgG, Fluorescein, Isotype Control, Novus Biologicals™
Equine Isotype Control Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196912
Catalog No. NBP196912
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IgG Isotype Control antibody specifically detects IgG in Equine samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
IgG
0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Thimerosal
immunoglobulin heavy constant gamma 1 (G1m marker)
IgG
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
Fluorescein
Lyophilized
Equine
Ion exchange chromatography
RUO
Isotype Control
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Fluorescein, anti-Equine IgG and anti-Equine Serum.
Equine