Learn More
anti-Horse IgG, HRP, Isotype Control, Novus Biologicals™
Equine Isotype Control Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196883
Catalog No. NBP196883
Add to cart
Description
Description
IgG Isotype Control antibody specifically detects IgG in Equine samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
IgG
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Gentamicin Sulfate
immunoglobulin heavy constant gamma 1 (G1m marker)
Equine
Ion exchange chromatography
RUO
Isotype Control
3500
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
HRP
Lyophilized
IGHG1
IgG
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Equine IgG and anti-Equine Serum.
Equine