Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Horse IgM, Isotype Control, Novus Biologicals™

Horse Isotype Control

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196970

Catalog No. NBP196970


Add to cart

Description

Description

IgM antibody specifically detects IgM in Equine samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

IgM
Unconjugated
Concentrated
Equine
Size Exclusion Chromatography
RUO
Isotype Control
3507
Equine
ELISA, Immunoprecipitation
0.1 M Tris Chloride, 0.5 M Sodium Chloride, pH 8.0. Stabilizer: None. with 0.1% Sodium Azide
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH
IgM
1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Equine Serum and anti-Equine IgM (mu chain specific). No reaction was observed against anti-Equine IgG F(c). Some light chain cross reactivity will occur with anti-Equine IgG.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only