Learn More
HOXA6, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human HOXA6 peptide using ARM Technology.
Supplier: Abnova Corporation H00003203K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
HOXA6 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
HOXA6 | |
HOXA6 | |
A synthetic peptide of human HOXA6 | |
RUO | |
3203 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
homeobox A6 | |
HOXA6 | |
HOX1, HOX1.2, HOX1B | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.