Learn More
HOXA9, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human HOXA9 peptide using ARM Technology.
Supplier: Abnova Corporation H00003205K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. This gene is highly similar to the abdominal-B (Abd-B) gene of Drosophila. A specific translocation event which causes a fusion between this gene and the NUP98 gene has been associated with myeloid leukemogenesis. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
HOXA9 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
HOXA9 | |
HOXA9 | |
A synthetic peptide of human HOXA9 | |
RUO | |
3205 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
homeobox A9 | |
HOXA9 | |
ABD-B, HOX1, HOX1.7, HOX1G, MGC1934 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.