Learn More
anti-HOXA9, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP224633SS
 View more versions of this product
Catalog No. NBP224633SS
Add to cart
Description
Description
HOXA9 Polyclonal antibody specifically detects HOXA9 in Human, Mouse, Rat, Chicken, Feline, Primate samples. It is validated for Western Blot.
Specifications
Specifications
HOXA9
0.5 mg/mL
Western Blot 1-3 ug/ml
P31269
ABD-B, homeo box A9, homeobox A9, Homeobox protein Hox-1G, homeobox protein Hox-A9, homeobox protein HOXA9, homeodomain protein HOXA9, HOX1, HOX1.7, HOX1GMGC1934
Rabbit
IgG
0.025 mg
Cancer
Primary
3205
Human, Mouse, Rat, Chicken, Feline, Primate
Western Blot
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Purified
HOXA9
A portion of amino acids 225-272 of human HOXA9 was used as the immunogen.
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
The amino acid sequence used as immunogen is 100% homologous in human, chimp, rhesus monkey, cat and horse, 92% homologous in dog and 66% homologous in mouse.