Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-HOXB4, Polyclonal, Novus Biologicals™
Manufacturer: Novus Biologicals NBP128475
Specifications
HOXB4 | |
0.5 mg/mL | |
Peptide ELISA Detection limit 1:32000 | |
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide | |
homeo box 2F, homeo box B4, homeobox B4, Homeobox protein Hox-2.6, Homeobox protein Hox-2F, homeobox protein Hox-B4, HOX-2.6, HOX2FHOX2 | |
HOXB4 | |
Peptide with sequence C-EYSQSDYLPSDH corresponding to internal region according to NP_076920.1. | |
Polyclonal | |
Immunogen affinity purified | |
RUO | |
Human,Mouse |
ELISA | |
Unconjugated | |
Affinity Purified | |
NP_076920.1 | |
3214 | |
Goat | |
IgG | |
Primary | |
0.1 mg | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only