Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXB4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224561
Description
HOXB4 Polyclonal specifically detects HOXB4 in Human, Mouse, Rat, Bovine, Feline, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HOXB4 | |
Polyclonal | |
Unconjugated | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
homeo box 2F, homeo box B4, homeobox B4, Homeobox protein Hox-2.6, Homeobox protein Hox-2F, homeobox protein Hox-B4, HOX-2.6, HOX2FHOX2 | |
Rabbit | |
Affinity Purified | |
RUO | |
3214 | |
Human, Mouse, Rat, Bovine, Feline, Primate | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
P17483 | |
HOXB4 | |
A portion of amino acids 100-150 of human HOXB4 was used as the immunogen. | |
0.1 mg | |
Primary | |
The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, chimp, cat, dogand horse. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction