Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXC11, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Supplier:  Thermo Scientific PA127700

Catalog No. PIPA127700

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis. Binds to a promoter element of the lactase-phlorizin hydrolase gene. This pro

PA1-27700 detects HOXC11 from human, mouse, and rat samples.PA1-27700 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27863 immunogen is a synthetic peptide corresponding to human Homeobox protein Hox-C11.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HOXC11
Polyclonal
Unconjugated
O43248, P31313
HOXC11
Synthetic peptide corresponding to human Homeobox protein Hox-C11.
50 μg
Primary
Human, Mouse, Rat
Antibody
IgY
Western Blot
1mg/mL
HOXC11
HOX3H, MGC4906
Chicken
Antigen affinity chromatography
RUO
3227
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit