Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXC6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525156

Catalog No. PIPA525156

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

This protein belongs to the homeobox family, members of which encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene, HOXC6, is one of several HOXC genes located in a cluster on chromosome 12. Three genes, HOXC5, HOXC4 and HOXC6, share a 5 ft. non-coding exon.
Specifications

Specifications

HOXC6
Polyclonal
PBS with 0.09% sodium azide
P09630
CP25, HHO.C8, HOX3, HOX3C, homeo box 3C, homeo box C6, homeo box C8 protein, homeobox protein CP25, homeobox protein HHO.C8, homeobox protein Hox-3C, homeobox protein Hox-C6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3223
Flow Cytometry, Western Blot
Unconjugated
HOXC6
Liquid
HOXC6
KLH conjugated synthetic peptide between 200-227 amino acids from the C-terminal region of human HOXC6
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit