Learn More
anti-HP1 alpha, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP224601SS
 View more versions of this product
Catalog No. NBP224601SS
Add to cart
Description
Description
HP1 alpha Polyclonal antibody specifically detects HP1 alpha in Human, Mouse, Canine, Primate samples. It is validated for Western Blot.
Specifications
Specifications
HP1 alpha
0.5 mg/mL
Western Blot 2-4 ug/ml
P45973
Antigen p25, chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, heterochromatin protein 1-alpha, HP1, HP1 alpha, HP1 alpha homolog, HP1A, HP1-ALPHA, HP1Hs alpha, HP1Hs-alpha
Rabbit
IgG
0.025 mg
Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing
Primary
23468
Human, Mouse, Canine, Primate
Western Blot
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Purified
CBX5
A portion of amino acids 150-200 of human CBX5 was used as the immunogen.
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
The amino acid sequence used as immunogen is 100% homologous in human, monkey, chimpanzee and chicken, and 94% homologous in horse, cow, mouse and rat.