Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HPCL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP214975
Description
HPCL Polyclonal specifically detects HPCL in Human, Mouse samples. It is validated for Western Blot.Specifications
HPCL | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
1600020H07Rik, 2,2-HPCL, 2-hydroxyacyl-CoA lyase 1, 2-hydroxyphytanoyl-CoA lyase2-hydroxyphytanol-CoA lyase, EC 4.1, HPCL, PHYH2FLJ55041, Phytanoyl-CoA 2-hydroxylase 2, phytanoyl-CoA hydroxylase 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
26061 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.36mg/mL | |
Western Blot 1:500-1:3000 | |
Q9UJ83 | |
HACL1 | |
Recombinant protein encompassing a sequence within the center region of human HACL1. The exact sequence is proprietary. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction