Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFTK1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525351

Catalog No. PIPA525351

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with rabbit based on sequence homology.

PFTK1 is a member of the CDC2 (MIM 116940)-related protein kinase family.
Specifications

Specifications

PFTK1
Polyclonal
Unconjugated
CDK14
Liquid
CDK14
KLH conjugated synthetic peptide between 1-30 amino acids from human hPFTK1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
O94921, O35495
PFTAIRE1, PFTK1, PFTAIRE protein kinase 1, cell division protein kinase 14, hPFTAIRE1, serine/threonine-protein kinase PFTAIRE-1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18647, 5218
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit