Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hPrp16 Mouse anti-Human, Unlabeled, Clone: 50, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611323

Catalog No. BDB611323


Add to cart

Description

Description

Splicing, the removal of introns from pre-mRNA, is mediated by spliceosomal complexes and occurs in two distinct catalytic steps. The first step involves cleavage of the 5 ft. exon and the production of a lariat intermediate. In the second step, the 3 ft.-splice site is cleaved and the exons are fused with concomitant release of the intron lariat. The spliceosome contains multiple snRNPs and a number of non-snRNP splicing factors. Four yeast proteins (Prp16p, Prp17p, Prp18p, and Slu7p) function exclusively in the second catalytic step. Human
homologs have been identified for Prp16p (hPrp16), Prp17p (hPrp17), and Prp18p (hPrp18). Prp16 is a DExD/DExH-box family RNA dependent ATPase, which can unwind RNA duplexes. hPrp16 is 41% identical to its yeast counterpart and chimeric yeast-human Prp16 can rescue a yeast Prp16 knockout strain. The non-conserved N-terminal region of hPrp16 is essential for viability, required for nuclear localization, and capable of binding to the splicesome during the second catalytic step. Thus, hPrp16 is a non-snRNP that is critical for spliceosomal function in the process of pre-mRNA splicing.

Host Species: Mouse
Clone: 50
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human hPrp16 1002-1203
Formula Weight [Chemical]: 140kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

hPrp16
50
Unconjugated
Murine
IgG1
RUO
Primary
Human
Immunofluorescence, Western Blot
250μg/mL
Affinity Purified
Human hPrp16 1002-1203
150μg
Cell Biology
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit