Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPS3 Goat anti-Human, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131349

Catalog No. PIPA131349


Add to cart

Description

Description

PA1-31349 detects HPS3 from human samples. PA1-31349 has been successfully used in Western blot applications. The PA1-31349 immunogen is: Synthetic peptide: PYLLYCSRKKPLT, corresponding to C terminal amino acids 992-1004 of Human HPS3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a protein containing a potential clathrin-binding motif, consensus dileucine signals, and tyrosine-based sorting signals for targeting to vesicles of lysosomal lineage. The encoded protein may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 3. Alternate splice variants exist, but their full length sequence has not been determined.
Specifications

Specifications

HPS3
Polyclonal
Unconjugated
HPS3
Liquid
HPS3
synthetic peptide PYLLYCSRKKPLT, corresponding to C terminal amino acids 992-1004 of Human HPS3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q969F9
DKFZp686F0413, FLJ22704, SUTAL
Goat
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
84343
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit