Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

v-Ha-ras Harvey rat sarcoma viral oncogene homolog, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human HRAS protein.

Manufacturer:  Abnova Corporation H00003265B01

Catalog No. 89-005-567


Add to cart

Description

Description

This gene belongs to the Ras oncogene family, whose members are related to the transforming genes of mammalian sarcoma retroviruses. The products encoded by these genes function in signal transduction pathways. These proteins can bind GTP and GDP, and they have intrinsic GTPase activity. This protein undergoes a continuous cycle of de- and re-palmitoylation, which regulates its rapid exchange between the plasma membrane and the Golgi apparatus. Mutations in this gene cause Costello syndrome, a disease characterized by increased growth at the prenatal stage, growth deficiency at the postnatal stage, predisposition to tumor formation, mental retardation, skin and musculoskeletal abnormalities, distinctive facial appearance and cardiovascular abnormalities. Defects in this gene are implicated in a variety of cancers, including bladder cancer, follicular thyroid cancer, and oral squamous cell carcinoma. Multiple transcript variants, which encode different isoforms, have been identified for this gene. [provided by RefSeq

Sequence: MTEYKLVVVGAGGVGKSALTIQLIQNHFVDEYDPTIEDSYRKQVVIDGETCLLDILDTAGQEEYSAMRDQYMRTGEGFLCVFAINNTKSFEDIHQYREQIKRVKDSDDVPMVLVGNKCDLAARTVESRQAQDLARSYGIPYIETSAKTRQGVEDAFYTLVREIRQHKLRKLNPPDESGPGCMSCKCVLS
Specifications

Specifications

v-Ha-ras Harvey rat sarcoma viral oncogene homolog
Unconjugated
No additive
NM_005343
C-BAS/HAS|C-H-RAS|C-HA-RAS1|CTLO|H-RASIDX|HAMSV|HRAS1|K-RAS|N-RAS|RASH1
Mouse
50 uL
Cell Cycle
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human HRAS protein.
HRAS
Serum
HRAS
HRAS (NP_005334, 1 a.a. ~ 189 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
3265
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit