Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRASLS3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525650

Catalog No. PIPA525650


Add to cart

Description

Description

PLA2G16 specifically catalyzes the release of fatty acids from phospholipids in adipose tissue. It also has a weak lysophospholipase activity. Tumor suppressor that may be involved in interferon-dependent cell death.
Specifications

Specifications

HRASLS3
Polyclonal
PBS with 0.09% sodium azide
P53816, Q8R3U1
AdPLA, H-REV107-1, HRASLS3, HREV107, HREV107-1, HREV107-3, HRSL3, Ca-independent phospholipase A1/2, H-rev 107 protein homolog, HRAS-like suppressor 1, HRAS-like suppressor 3, adipose-specific PLA2, adipose-specific phospholipase A2, group XVI phospholipase A2, renal carcinoma antigen NY-REN-65
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11145, 225845
Flow Cytometry, Western Blot
Unconjugated
PLA2G16
Liquid
PLA2G16
KLH conjugated synthetic peptide between 104-132 amino acids from the C-terminal region of human HRASLS3
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit