Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRD1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524593

Catalog No. PIPA524593


Add to cart

Description

Description

This antibody is predicted to react with mouse and Xenopus based on sequence homology.

HRD1 encodes a protein involved in endoplasmic reticulum (ER)-associated degradation. The encoded protein removes unfolded proteins, accumulated during ER stress, by retrograde transport to the cytosol from the ER. This protein also uses the ubiquitin-proteasome system for additional degradation of unfolded proteins. This gene and the mitochondrial ribosomal protein L49 gene use in their respective 3 ft. UTRs some of the same genomic sequence.
Specifications

Specifications

HRD1
Polyclonal
PBS with 0.09% sodium azide
Q86TM6
HRD1, E3 ubiquitin-protein ligase synoviolin
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84447
Western Blot
Unconjugated
SYVN1
Liquid
SYVN1
KLH conjugated synthetic peptide between 106-138 amino acids from the N-terminal region of human HRD1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit