Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HS1 Mouse anti-Human, Unlabeled, Clone: 9, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610540

 View more versions of this product

Catalog No. BDB610540


Add to cart

Description

Description

HS1 (lck-binding protein 1 (lckBP1)) is a protein of 486 amino acids with homology to cytoplasmic tyrosine kinases. The protein contains an SH3 domain and two proline-rich regions, that may be the binding site for lck. Co- mmunoprecipitation experiments demonstrated a tight association between HS1 and lck. The SH3 domain of lck is necessary for this interaction. In addition, HS1 contains tandem repeat sequence of seven amino acids which is a substrate for the tyrosine kinase src. This sequence is also found in the protein cortactin. Therefore, HS1, like cortactin, may be an important adaptor that is critical for the regulation of cellular tyrosine kinases.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

HS1
9
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.