Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HS2ST1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514675

Catalog No. PIPA514675

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. Heparan sulfate 2-O-sulfotransferase is a member of the heparan sulfate biosynthetic enzyme family. This family member transfers sulfate to the 2 position of the iduronic acid residue of heparan sulfate. The disruption of the HS2ST1 gene resulted in no kidney formation in knockout embryonic mice, indicating that the absence of this enzyme may interfere with the signaling required for kidney formation.
Specifications

Specifications

HS2ST1
Polyclonal
PBS with 0.09% sodium azide
Q7LGA3
Heparan sulfate 2-O-sulfotransferase 1; 2-O-sulfotransferase; 2OST; HS2ST1; HS2ST; KIAA0448
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9653
Western Blot
Unconjugated
HS2ST1
Liquid
HS2ST1
KLH conjugated synthetic peptide between 16-45 amino acids from the N-terminal region of human HS2ST1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit