Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

HSD17B2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525049

Catalog No. PIPA525049



Description

Description

HSD17B2 is capable of catalyzing the interconversion of testosterone and androstenedione, as well as estradiol and estrone.HSD17B2 also has 20-alpha-HSD activity. HSD17B2 uses NADH while EDH17B3 uses NADPH.
Specifications

Specifications

HSD17B2
Polyclonal
PBS with 0.09% sodium azide
P37059
EDH17B2, HSD17, SDR9C2, 17-beta-HSD 2, 17-beta-hydroxysteroid dehydrogenase type 2, 20 alpha-hydroxysteroid dehydrogenase, 20-alpha-HSD, E2DH, estradiol 17-beta-dehydrogenase 2, microsomal 17-beta-hydroxysteroid dehydrogenase, short chain dehydrogenase/reductase family 9C, member 2, tes
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3294
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
HSD17B2
Liquid
HSD17B2
KLH conjugated synthetic peptide between 265-294 amino acids from the central region of human HSD17B2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit