Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526524

Catalog No. PIPA526524

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This isoform of 17 beta-hydroxysteroid dehydrogenase is expressed predomitly in the testis and catalyzes the conversion of androstenedione to testosterone. It preferentially uses NADP as cofactor. Deficiency can result in male pseudohermaphroditism with gynecomastia.
Specifications

Specifications

HSD17B3
Polyclonal
PBS with 0.09% sodium azide
P37058
EDH17B3, SDR12C2, 17-beta-HSD 3, 17-beta-HSD3, 17-beta-hydroxysteroid dehydrogenase type 3, short chain dehydrogenase/reductase family 12C, member 2, testicular 17-beta-hydroxysteroid dehydrogenase, testosterone 17-beta-dehydrogenase 3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3293
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
HSD17B3
Liquid
HSD17B3
KLH conjugated synthetic peptide between 89-118 amino acids from the central region of human HSD17B3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit