Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp110 Mouse, Unlabeled, Clone: 21, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610510

Catalog No. BDB610510


Add to cart

Description

Description

With the exception of Hsp110, the stress or heat shock proteins have been thoroughly characterized. Hsp110 has been reported to be expressed in most tissues. Like other stress proteins, Hsp110 is induced by heat shock and its induction is associated with thermotolerance. The sequence of Hsp110 reportedly bears similarity to several Hsp70-related proteins that have been termed Hsp110/SSE. The carboxy-terminal domains of these proteins and Hsp110 contain a putative peptide-binding site. These proteins also contain six highly conserved regions found in the same progression, as well as five conserved ATP-binding motifs.
Specifications

Specifications

Hsp110
21
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Heat Shock Protein-110
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit