Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HSP60, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB10056469

 View more versions of this product

Catalog No. NB10056469


Add to cart

Description

Description

HSP60 Polyclonal antibody specifically detects HSP60 in Human, Bovine, Canine, Chicken, Primate samples. It is validated for Western Blot.
Specifications

Specifications

HSP60
0.5 mg/mL
Western Blot 0.2-1 ug/ml
P10809
60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant)
Rabbit
IgG
0.1 mg
Apoptosis, Membrane Trafficking and Chaperones
Primary
3329
Human, Bovine, Canine, Chicken, Primate
Western Blot
Unconjugated
PBS containing 0.2% gelatin with 0.05% Sodium Azide
Purified
HSPD1
A portion of amino acids 350-400 of human Hsp60 was used as the immunogen.
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
The amino acid sequence used as immunogen is 100% homologous in a broad range of species including human, dog, mouse, rat, bovine, chicken, chimp, monkey, and orangutan.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only