Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA1B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA529605

Catalog No. PIPA529605


Add to cart

Description

Description

PA5-29606 has successfully been used to detect CD29 in IF, IHC and WB in human and horse. The PA5-29606 immunogen is recombit fragment corresponding to a region within amino acids 128 and 410 of Human Integrin beta 1.

This intronless gene encodes a 70kDa heat shock protein which is a member of the heat shock protein 70 family. In conjuction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.
Specifications

Specifications

HSPA1B
Polyclonal
Unconjugated
HSPA1B
Liquid
HSPA1B
Recombit protein fragment corresponding to a region within amino acids 190 and 423 of Human HSP70 1B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
P0DMV9
HSP70-1; HSP70-1A; HSP70I; HSP72; HSPA1; HSP70-1/HSP70-2; HSP70.1/HSP70.2; dnaK-type molecular chaperone HSP70-1; heat shock 70 kDa protein 1/2; heat shock 70 kDa protein 1A/1B; heat shock 70kD protein 1A; heat shock-induced protein
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3304
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit