anti-HSP70/HSPA1A, Clone: 242707, R&D Systems

Manufacturer: R&D Systems A400

 View more versions of this product

Catalog No. A400

  Check Availability
Add to Cart

    Specifications

    Antigen HSP70/HSPA1A
    Format Purified
    Host Species Mouse
    Immunogen Met1-Asp641
    Isotype IgG2a
    Purification Method Protein A or G purified from hybridoma culture supernatant
    Quantity 50μg
    Regulatory Status RUO
    Primary or Secondary Primary
    Monoclonal or Polyclonal Monoclonal
    Clone 242707
    Conjugate Unlabeled
    Cross Reactivity Human,Mouse,Rat

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit