Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP70/HSPA1A Rabbit anti-Human, Rat, DyLight 550, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP177456R

Catalog No. NBP177456R


Add to cart

Description

Description

HSP70/HSPA1A Polyclonal antibody specifically detects HSP70/HSPA1A in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

HSP70/HSPA1A
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
HSPA1A
A synthetic peptide made to a C-terminus portion of the human Hsp70 protein (between residues 600-641) [UniProt # P08107]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
P08107
dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B
Rabbit
IgG
0.1 ml
Cancer, Cellular Markers, Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific
Primary
3303
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only