Learn More
HSP90 beta Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA527410
Catalog No. PIPA527410
Add to cart
Description
Description
PA5-27410 targets HSP90 beta in IF, IHC (P), and WB applications and shows reactivity with Human, rat, and mouse samples. The PA5-27410 immunogen is recombit fragment corresponding to a region within amino acids 207 and 549 of Human HSP90 beta. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Heat shock proteins (HSP) are expressed in response to various biological stresses, including heat. HSP90 is a 90 kDa protein that is induced under stress conditions, but is also one of the most abundant cellular proteins found under non-stress conditions. HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. HSP90 has been found to be associated with a number of other intracellular proteins, including steroid receptors, actin, tubulin, aryl hydrocarbon (Ah) receptor, and some kinases.
Specifications
Specifications
HSP90 beta
Polyclonal
Unconjugated
HSP90AB1
Liquid
HSP90AB1
Recombit fragment corresponding to a region within amino acids 207 and 549 of Human HSP90 beta
Protein A
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
5.43 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P08238, P11499, P34058
D6S182; HSP90-BETA; HSP90B; HSPC2; HSPCB; 90-kda heat shock protein beta HSP90 beta; HSP 84; HSP 90; HSP84; heat shock 84 kDa; heat shock 90kD protein 1; beta; heat shock 90kDa protein 1; beta; heat shock protein HSP 90-beta; heat shock protein beta
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
15516, 301252, 3326